Learn More
Apexbio Technology LLC Madecassic acid(Synonyms: Madecassic acid, Madecassate, Madecassic Acid, Terminolic acid, 6-Hydroxyasiatic acid), 10mM (in 1mL DMSO), CAS: 18449-41-7.
SDP
Catalog No. 5000061129
Encompass_Preferred
Madecassic acid (CAS 18449-41-7) is a bioactive triterpenoid isolated from the whole herb of Centella asiatica It exhibits its biological activity primarily through modulating inflammatory pathways stimulating collagen synthesis and promoting wound healing Recent studies indicate that madecassic acid can suppress pro-inflammatory mediators and cytokines enhancing its relevance in studies of inflammation-mediated conditions and tissue regeneration Its pharmacological properties make it valuable for biomedical research focused on dermatological healing processes and inflammatory disorders Store compound in a dry cool and sealed environment
Catalog No. 50-000-61129 Supplier Apexbio Technology LLC Supplier No. N23555.1
Add to Cart
Add to Cart
Madecassic acid (CAS 18449-41-7) is a bioactive triterpenoid isolated from the whole herb of Centella asiatica It exhibits its biological activity primarily through modulating inflammatory pathways stimulating collagen synthesis and promoting wound healing Recent studies indicate that madecassic acid can suppress pro-inflammatory mediators and cytokines enhancing its relevance in studies of inflammation-mediated conditions and tissue regeneration Its pharmacological properties make it valuable for biomedical research focused on dermatological healing processes and inflammatory disorders Store compound in a dry cool and sealed environment