Learn More
Medchemexpress LLC Madecassic acid (Standard) | 18449-41-7 | 504.70 | 25 MG
SDP
Catalog No. 5000389322
Encompass_Preferred
Madecassic acid is an analytical and reference standard isolated from Centella asiatica (Umbelliferae). It is intended for research and analytical applications, and exhibits anti-inflammatory properties by inhibiting iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6 through the downregulation of NF-κB activation in RAW 264.7 macrophage cells.
  • Analytical and reference standard
  • Isolated from Centella asiatica (Umbelliferae)
  • Exhibits anti-inflammatory properties by inhibiting iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS
Catalog No. 50-003-89322 Supplier Medchemexpress LLC Supplier No. 25MGHYN0569R
Add to Cart
Add to Cart
Madecassic acid is an analytical and reference standard isolated from Centella asiatica (Umbelliferae). It is intended for research and analytical applications, and exhibits anti-inflammatory properties by inhibiting iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6 through the downregulation of NF-κB activation in RAW 264.7 macrophage cells.
  • Analytical and reference standard
  • Isolated from Centella asiatica (Umbelliferae)
  • Exhibits anti-inflammatory properties by inhibiting iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS