Learn More
Medchemexpress LLC Madecassic acid (Standard) | 18449-41-7 | 504.70 | 5 MG
SDP
Catalog No. 5000388895
Encompass_Preferred
Madecassic acid (Standard) is the analytical standard of Madecassic acid, intended for research and analytical applications. It is isolated from *Centella asiatica* (Umbelliferae) and exhibits anti-inflammatory properties through the downregulation of NF-κB activation in RAW 264.7 macrophage cells.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-003-88895 Supplier Medchemexpress LLC Supplier No. HYN0569R5MG
Madecassic acid (Standard) is the analytical standard of Madecassic acid, intended for research and analytical applications. It is isolated from *Centella asiatica* (Umbelliferae) and exhibits anti-inflammatory properties through the downregulation of NF-κB activation in RAW 264.7 macrophage cells.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications