Learn More
Medchemexpress LLC Madecassic acid (Standard) | 18449-41-7 | 504.70 | 50 MG
SDP
Catalog No. 5000388894
Encompass_Preferred
Madecassic acid (Standard) is an analytical standard intended for research and analytical applications. It is isolated from *Centella asiatica* and possesses anti-inflammatory properties through the inhibition of iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6 via downregulation of NF-κB activation in RAW 264.7 macrophage cells. This compound serves as a reference standard, commonly used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Isolated from *Centella asiatica*.
  • Exhibits anti-inflammatory properties.
  • Inhibits iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6.
  • Downregulates NF-κB activation in macrophage cells.
  • Used in HPLC, GC, and MS research.
Catalog No. 50-003-88894 Supplier Medchemexpress LLC Supplier No. HYN0569R50MG
Madecassic acid (Standard) is an analytical standard intended for research and analytical applications. It is isolated from *Centella asiatica* and possesses anti-inflammatory properties through the inhibition of iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6 via downregulation of NF-κB activation in RAW 264.7 macrophage cells. This compound serves as a reference standard, commonly used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Isolated from *Centella asiatica*.
  • Exhibits anti-inflammatory properties.
  • Inhibits iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6.
  • Downregulates NF-κB activation in macrophage cells.
  • Used in HPLC, GC, and MS research.