Learn More
Cayman Chemical Maduramicin (ammonium salt), Purity: ≥95%, CAS: 84878-61-5, 100 mg
SDP
Catalog No. 502950801
Encompass_Preferred
A polyether ionophore antibiotic; selectively binds to monovalent cations over divalent cations; active against Gram-positive bacteria (MICs = 0.9-125 µg/ml); reduces the viability of asexual and gametocyte P. falciparum parasites (EC50s = 1.3 and 14.8 nM, respectively); reduces P. berghei transmission in a mouse model of malaria at 4 and 8 mg/kg; reduces the severity of lesions and completely protects chicks against mortality in a model of E. tenella-induced coccidiosis at 5-10 ppm in the diet
Catalog No. 50-295-0801 Supplier Cayman Chemical Supplier No. 34270100 MG
A polyether ionophore antibiotic; selectively binds to monovalent cations over divalent cations; active against Gram-positive bacteria (MICs = 0.9-125 µg/ml); reduces the viability of asexual and gametocyte P. falciparum parasites (EC50s = 1.3 and 14.8 nM, respectively); reduces P. berghei transmission in a mouse model of malaria at 4 and 8 mg/kg; reduces the severity of lesions and completely protects chicks against mortality in a model of E. tenella-induced coccidiosis at 5-10 ppm in the diet