Learn More
Selleck Chemical LLC Maduramycin Ammonium S5094-25mg
SDP
Catalog No. 503112378
Encompass_Preferred
Maduramicin which could be isolated from the actinomycete Actinomadura rubra is an antiprotozoal agent used in veterinary medicine to prevent coccidiosis
Catalog No. 50-311-2378 Supplier Selleck Chemical LLC Supplier No. S509425MG
Add to Cart
Add to Cart
Maduramicin which could be isolated from the actinomycete Actinomadura rubra is an antiprotozoal agent used in veterinary medicine to prevent coccidiosis