Learn More
Mopec Maestro 1/2 HP Disposal
SDP
Catalog No. 23771089 Shop All Mopec Products
Change view
Click to view available options
Includes:
1/2 HP Disposal
Catalog No. Includes
23-771-089 1/2 HP Disposal
1 options
Catalog No. 23-771-089 Supplier Mopec Supplier No. MB1024
Add to Cart
Add to Cart
Upgrade option for legacy grossing stations without a disposal
  • Upgrade option for legacy grossing stations that were ordered without a disposal
  • Now standard in all Maestro Encore grossing stations
  • Polished stainless steel
  • 1/2 Horsepower
  • 2600 RPM
  • Compatible with all Maestro and Maestro Encore grossing stations
Specifications
Product Type Grossing Station
Material Stainless Steel
Includes 1/2 HP Disposal