Learn More
Mopec Maestro Coupler Base Magnifier
SDP
Catalog No. 23771091 Shop All Mopec Products
Change view
Click to view available options
Includes:
Base Magnifier
Catalog No. Includes
23-771-091 Base Magnifier
1 options
Catalog No. 23-771-091 Supplier Mopec Supplier No. MB1029
Add to Cart
Add to Cart
Designed to mount to the shelf of your grossing station with 2X magnification
  • Designed to mount to the shelf of your Maestro or Maestro Encore grossing station to provide 2X magnification
  • Includes a coupler base for quick-release swivel mounting
  • Glass 2X magnifier
  • Vinyl-coated flex arm
  • Magnifier: 4 in.
  • Flexible Arm: 24 in.
  • Compatible with all Maestro and Maestro Encore grossing stations
Specifications
Product Type Grossing Station
Includes Base Magnifier