Learn More
Mopec Maestro Formalin Dispensing Tap for FP100 Formalin Pumping Station
SDP
Catalog No. 21010376 Shop All Mopec Products
Change view
Click to view available options
Material:
Chrome-plated Brass
Catalog No. Material
21-010-376 Chrome-plated Brass
1 options
Catalog No. 21-010-376 Supplier Mopec Supplier No. MB1037
Add to Cart
Add to Cart
Maestro Formalin Dispensing Tap For FP100 Formalin Pumping Station provides a simple, reliable solution for dispensing formalin from a central FP100 system. Made of chrome-plated brass, this tap is compatible with all Maestro stations and is designed for use with external formalin pumping systems only.
Construction
  • Designed to be used on Maestro and Maestro Encore grossing stations in conjunction with a centrally-located FP100 Formalin Pumping Station
  • Includes (1) dispensing tap
Materials
  • Chrome-plated brass
Compatibility
  • Compatible with all Maestro and Maestro Encore grossing stations
  • To be used in conjunction with the FP100 Formalin Pumping Station
NOTE :
  • This option is different than the MB1040 Formalin Dispensing System and the MB1043 Formalin Dispensing and Collection System, which have an internal tank within the grossing station for pumping formalin.
  • This option must be used with an external formalin pumping station, such as the FP100.
Specifications
Product Type Grossing Station
Material Chrome-plated Brass