Learn More
Mopec Maestro Hand Spray Rinse - Right
SDP
Catalog No. 21010374 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Grossing Station
Catalog No. Product Type
21-010-374 Grossing Station
1 options
This item is not returnable. View return policy
Catalog No. 21-010-374 Supplier Mopec Supplier No. MB1032R
Add to Cart
Add to Cart
This item is not returnable. View return policy
Maestro Hand Spray Rinse provides a convenient and efficient way to rinse down grossing station work surfaces. Included are a 60 in. hose, chrome-plated brass sprayer, and flow control fixture with a vacuum breaker. Constructed of durable stainless steel, it's now standard in all Maestro Encore stations.
Construction
  • Designed for customers to rinse off the workstation of their grossing station
  • Includes (1) sprayer, (2) 60 in. hose, flow control fixture, plumbing and fittings
  • Placed on the opposite side of the unit's sink
  • Now standard in all Maestro Encore grossing stations
Dimensions
  • 60 in. hose
Materials
  • Hand sprayer with soft spray : chrome-plated brass
  • Sheetmetal : 304 stainless steel construction with a polished No. 4 finish
  • Hardware : 304 stainless steel
  • Plumbing lines and fittings : brass, pex, and stainless steel
Performance
  • Mixing valve/flow control fixture with vacuum breaker
  • Backflow preventers on both hot and cold-water lines
Options and Accessories
  • MB1032L : designed for left-hand Maestro grossing stations
  • MB1032R : designed for right-hand Maestro grossing stations
Compatibility
  • Compatible with all Maestro grossing stations that do not currently have this option
NOTE : For stations adding this option where it had not previously existed, it would include a new endcap to the work surface with a hole for the hand spray to retract inside the station.
Specifications
Product Type Grossing Station
Color Red