Learn More
Mopec Maestro Red GCFI And USB Outlets
SDP
Catalog No. 21010372 Shop All Mopec Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
21-010-372 Red
1 options
Catalog No. 21-010-372 Supplier Mopec Supplier No. MB1031
Add to Cart
Add to Cart
Maestro Red GFCI and USB Outlets are designed to clearly indicate connection to a backup power source. This kit includes a hospital-grade GFCI outlet and a USB-equipped receptacle, offering power indication, 15/20 amp feed-through, and 125V. Compatible with all Maestro stations.
Construction
  • Designed for customers who need red outlets to indicate they are connected to a backup power source
  • Includes one heavy-duty, hospital-grade, tamper-resistant, duplex receptacle outlet with two type-A USB ports; one extra heavy-duty, hospital grade, self-test GFCI duplex receptacle outlet; and wall plate
Performance
  • Features power indication, 15 amp and 20 amp feed-through, and 125 volt
  • Receptacle outlet with USB is located inside the Maestro column behind touchscreen
  • GFCI receptacle outlet is located on the outside of the Maestro column on the opposite side of sink
Compatibility
  • Compatible with all Maestro and Maestro Encore grossing stations
  • Compatible with back or side wiring, NEMA 5-15R, 2-pole, 3-wire, and self-grounding.
Specifications
Product Type Grossing Station
Color Red