Learn More
Mopec Maestro T-Handle Drain Plug
SDP
Catalog No. 23987018 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Drain Plug
Catalog No. For Use With (Equipment)
23-987-018 Drain Plug
1 options
Catalog No. 23-987-018 Supplier Mopec Supplier No. AX101
Add to Cart
Add to Cart
Designed to be used in smaller sinks that are built into grid plates and for center cup.
  • Designed to be used in smaller sinks that are built into grid plates and for center cup sinks on the Maestro Encore grossing stations
  • The long T-shaped handle keeps the user's hands out of harms way in the sink
  • 304 stainless steel handle and hardware
  • SBR rubber drain plug
  • Fits small 1.125 diameter drain hole used in smaller sinks
  • Does not fit standard sink drains or garbage disposal of Maestro Encore grossing stations
Specifications
Product Type Grossing Station
For Use With (Equipment) Drain Plug