Learn More
Aviva Systems Biology Corp MAF Antibody - N-terminal region (ARP38607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAF. Alternate names: MGC71685, c-MAFApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56725