Learn More
Aviva Systems Biology Corp Maf1 Antibody - N-terminal region (ARP50710_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50710_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Maf1. It was validated on Western Blot. . Alternate names: 1110068E11Rik, AU042856ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-530