Learn More
Aviva Systems Biology Corp Mafa Antibody - middle region (ARP37615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mafa. It was validated on Western Blot. Mafa,v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian),RIPE3b1; Mafa,transcription factor MafA,transcription factor MafA,Mafa homolog,pancreatic beta-cell-specific transcriptional activator,V-maf musculoaponeurotic fibrosarcoma oncogene homolog AThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: RIPE3b1ApplicationWB. Species reactivity: Cow, Mouse, Rat
Catalog No. 50-284-671