Learn More
Aviva Systems Biology Corp MAFB Antibody - N-terminal region (ARP33683_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33683_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MAFB. Alternate names: KRML, MGC43127ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52917