Learn More
Aviva Systems Biology Corp MAFB Antibody - C-terminal region (ARP58149_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58149_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAFB. Alternate names: KRML, MGC43127ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54077