Learn More
Aviva Systems Biology Corp MAFB Antibody - middle region (ARP58150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MAFB. Alternate names: KRML, MGC43127ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Zebrafish
Catalog No. 50-286-444