Learn More
Apexbio Technology LLC Mafenide Acetate 13009-99-9 200mg
SDP
Catalog No. 5000096394
Encompass_Preferred
Mafenide Acetate (CAS 13009-99-9) is a small-molecule inhibitor targeting bacterial nucleotide biosynthesis pathways It is designed to inhibit nucleic acid production by interfering with microbial folic acid synthesis Mafenide Acetate exerts its biological activity primarily through competitive inhibition of para-aminobenzoic acid disrupting folic acid synthesis essential for bacterial growth In in vitro studies Mafenide Acetate demonstrates antibacterial activity against selected Gram-positive and Gram-negative bacteria including Pseudomonas aeruginosa typically showing inhibitory effects with IC50 values in the micromolar range Based on these pharmacological properties Mafenide Acetate holds research potential in microbial infection control and wound care particularly for studies involving burn-associated bacterial infections
Catalog No. 50-000-96394 Supplier Apexbio Technology LLC Supplier No. B3480200
Mafenide Acetate (CAS 13009-99-9) is a small-molecule inhibitor targeting bacterial nucleotide biosynthesis pathways It is designed to inhibit nucleic acid production by interfering with microbial folic acid synthesis Mafenide Acetate exerts its biological activity primarily through competitive inhibition of para-aminobenzoic acid disrupting folic acid synthesis essential for bacterial growth In in vitro studies Mafenide Acetate demonstrates antibacterial activity against selected Gram-positive and Gram-negative bacteria including Pseudomonas aeruginosa typically showing inhibitory effects with IC50 values in the micromolar range Based on these pharmacological properties Mafenide Acetate holds research potential in microbial infection control and wound care particularly for studies involving burn-associated bacterial infections