Learn More
Medchemexpress LLC Mafenide Acetate | 13009-99-9 | 99.3% | 1 ML
SDP
Catalog No. 5000408282
Encompass_Preferred
Mafenide Acetate is a potent sulfonamide antimicrobial agent that shows antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*. It also demonstrates antifungal activity against filamentous fungi like *Lichtheimia* and *Aspergillus flavus*. This compound can be used in research related to skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent
  • Exhibits antibacterial activity against Staphylococcus aureus and Pseudomonas aeruginosa
  • Shows antifungal activity against filamentous fungi (e.g., Lichtheimia and Aspergillus flavus)
  • Can be used in research for skin grafts on burn wounds
  • Can be used in research for post-traumatic invasive fungal infections
  • Can be used in research for bacterially contaminated wounds
Catalog No. 50-004-08282 Supplier Medchemexpress LLC Supplier No. HYB0614A10MM/1ML
Mafenide Acetate is a potent sulfonamide antimicrobial agent that shows antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*. It also demonstrates antifungal activity against filamentous fungi like *Lichtheimia* and *Aspergillus flavus*. This compound can be used in research related to skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent
  • Exhibits antibacterial activity against Staphylococcus aureus and Pseudomonas aeruginosa
  • Shows antifungal activity against filamentous fungi (e.g., Lichtheimia and Aspergillus flavus)
  • Can be used in research for skin grafts on burn wounds
  • Can be used in research for post-traumatic invasive fungal infections
  • Can be used in research for bacterially contaminated wounds