Learn More
Medchemexpress LLC Mafenide Acetate | 13009-99-9 | 99.3% | 100 MG
SDP
Catalog No. 5000405910
Encompass_Preferred
Mafenide Acetate is a potent sulfonamide antimicrobial agent that exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*, as well as antifungal activity against filamentous fungi like *Lichtheimia* and *Aspergillus flavus*. It is useful for research concerning skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent.
  • Exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*.
  • Shows antifungal activity against filamentous fungi (e.g., *Lichtheimia* and *Aspergillus flavus*).
  • Useful for research on skin grafts on burn wounds.
  • Applicable in research for post-traumatic invasive fungal infections.
  • Suitable for research on bacterially contaminated wounds.
Catalog No. 50-004-05910 Supplier Medchemexpress LLC Supplier No. HYB0614A100MG
Mafenide Acetate is a potent sulfonamide antimicrobial agent that exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*, as well as antifungal activity against filamentous fungi like *Lichtheimia* and *Aspergillus flavus*. It is useful for research concerning skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent.
  • Exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*.
  • Shows antifungal activity against filamentous fungi (e.g., *Lichtheimia* and *Aspergillus flavus*).
  • Useful for research on skin grafts on burn wounds.
  • Applicable in research for post-traumatic invasive fungal infections.
  • Suitable for research on bacterially contaminated wounds.