Learn More
Medchemexpress LLC Mafenide (Acetate) | 13009-99-9 | 99.3% | 50 MG
SDP
Catalog No. 5000409038
Encompass_Preferred
Mafenide Acetate is a potent sulfonamide antimicrobial agent that demonstrates antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*, and antifungal activity against filamentous fungi (e.g., *Lichtheimia* and *Aspergillus flavus*). It is used in research for skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent
  • Exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*
  • Shows antifungal activity against filamentous fungi
  • Used in research for skin grafts on burn wounds
  • Applicable for post-traumatic invasive fungal infections
  • Used in research for bacterially contaminated wounds
Catalog No. 50-004-09038 Supplier Medchemexpress LLC Supplier No. HYB0614A50MG
Mafenide Acetate is a potent sulfonamide antimicrobial agent that demonstrates antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*, and antifungal activity against filamentous fungi (e.g., *Lichtheimia* and *Aspergillus flavus*). It is used in research for skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent
  • Exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*
  • Shows antifungal activity against filamentous fungi
  • Used in research for skin grafts on burn wounds
  • Applicable for post-traumatic invasive fungal infections
  • Used in research for bacterially contaminated wounds