Learn More
Medchemexpress LLC Mafenide Acetate | 13009-99-9 | 99.3% | 500 MG
SDP
Catalog No. 5000406842
Encompass_Preferred
Mafenide Acetate is a potent sulfonamide antimicrobial agent. It exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*, and also shows antifungal activity against filamentous fungi like *Lichtheimia* and *Aspergillus flavus*. It is suitable for research related to skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent
  • Exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*
  • Shows antifungal activity against filamentous fungi
  • Used in research for skin grafts on burn wounds
  • Applicable for post-traumatic invasive fungal infections research
  • Useful for bacterially contaminated wounds research
Catalog No. 50-004-06842 Supplier Medchemexpress LLC Supplier No. HYB0614A500MG
Mafenide Acetate is a potent sulfonamide antimicrobial agent. It exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*, and also shows antifungal activity against filamentous fungi like *Lichtheimia* and *Aspergillus flavus*. It is suitable for research related to skin grafts on burn wounds, post-traumatic invasive fungal infections, and bacterially contaminated wounds.
  • Potent sulfonamide antimicrobial agent
  • Exhibits antibacterial activity against *Staphylococcus aureus* and *Pseudomonas aeruginosa*
  • Shows antifungal activity against filamentous fungi
  • Used in research for skin grafts on burn wounds
  • Applicable for post-traumatic invasive fungal infections research
  • Useful for bacterially contaminated wounds research