Learn More
Medchemexpress LLC HY-B0614A 200mg Medchemexpress, Mafenide (Acetate) CAS:13009-99-9 Purity:>98%
SDP
Catalog No. 502082620
Encompass_Preferred
Medchemexpress, HY-B0614A 200mg Mafenide (Acetate) CAS:13009-99-9 Mafenide Acetate is an effective sulfonamide-type antimicrobial agent used for burn wounds. Mafenide Acetate shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2620 Supplier Medchemexpress LLC Supplier No. HYB0614A200MG
Medchemexpress, HY-B0614A 200mg Mafenide (Acetate) CAS:13009-99-9 Mafenide Acetate is an effective sulfonamide-type antimicrobial agent used for burn wounds. Mafenide Acetate shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.