Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mafenide hydrochloride | 138-37-4 | 99.97% | 100 MG
SDP

Supplier:  Medchemexpress LLC HYB0614B100MG

Encompass_Preferred

Mafenide hydrochloride is an effective sulfonamide-type antimicrobial agent used for burn wounds. It exhibits activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, by inhibiting nucleotide synthesis. It is a white to off-white solid with a purity of 99.97% and a molecular weight of 222.69.

  • Effective sulfonamide-type antimicrobial agent
  • Active against Gram-positive and Gram-negative organisms
  • Inhibits nucleotide synthesis
  • Purity of 99.97%
  • Soluble in DMSO (125 mg/mL)
  • Recommended storage at 4°C in sealed container, away from moisture

Catalog No. 50-004-07752


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.