Learn More
Medchemexpress LLC Mafenide hydrochloride | 138-37-4 | MFCD00013005 | 99.97% | 50 MG
SDP
Catalog No. 5000408308
Encompass_Preferred
Mafenide hydrochloride is an effective sulfonamide-type antimicrobial agent used for burn wounds. It shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis.
  • Effective sulfonamide-type antimicrobial agent
  • Used for burn wounds
  • Active against Gram-positive organisms
  • Active against Gram-negative organisms
  • Effective against Pseudomonas aeruginosa
  • Inhibits nucleotide synthesis
  • Regulatory Status: RUO
  • Ships at room temperature
  • Store at 4°C, sealed, and away from moisture
Catalog No. 50-004-08308 Supplier Medchemexpress LLC Supplier No. HYB0614B50MG
Mafenide hydrochloride is an effective sulfonamide-type antimicrobial agent used for burn wounds. It shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis.
  • Effective sulfonamide-type antimicrobial agent
  • Used for burn wounds
  • Active against Gram-positive organisms
  • Active against Gram-negative organisms
  • Effective against Pseudomonas aeruginosa
  • Inhibits nucleotide synthesis
  • Regulatory Status: RUO
  • Ships at room temperature
  • Store at 4°C, sealed, and away from moisture