Learn More
Aviva Systems Biology Corp MAFG Antibody - middle region (ARP32691_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32691_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAFG. Alternate names: MGC13090, MGC20149, hMAFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54709