Learn More
Aviva Systems Biology Corp MAFK Antibody - N-terminal region (ARP47559_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47559_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAFK. Alternate names: FLJ32205, MGC71717, NFE2U, P18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59587