Learn More
Enzo Life Sciences Methyl Arachidonyl Fluorophosphonate (25 mg). CAS: 188404-10-6
SDP
Catalog No. 502012168
Encompass_Preferred
Selective, active-site directed, irreversible inhibitor of cPLA2 (IC50=5µM) and Ca2+-independent iPLA2 (IC50=0.5µM). Potent, irreversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50=2.5nM), 800 -fold more potent than AACOCF3 (BML-ST335)). Irriversible CB1 receptor antagonist in rat brain membrane (IC50=20nM). Alternative name: MAFP, Methylphosphonofluoridic acid 5,8,11,14-eicosatetraenyl ester. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (50mg/ml) or DMSO (50mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-2168 Supplier Enzo Life Sciences Supplier No. BMLST3600025
Selective, active-site directed, irreversible inhibitor of cPLA2 (IC50=5µM) and Ca2+-independent iPLA2 (IC50=0.5µM). Potent, irreversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50=2.5nM), 800 -fold more potent than AACOCF3 (BML-ST335)). Irriversible CB1 receptor antagonist in rat brain membrane (IC50=20nM). Alternative name: MAFP, Methylphosphonofluoridic acid 5,8,11,14-eicosatetraenyl ester. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (50mg/ml) or DMSO (50mg/ml). Long Term Storage: -80°C.