Learn More
V&P Scientific Inc MAGNETIC RACK, 8 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume
Catalog No. NC2137756
Encompass
MAGNETIC BEAD SEPARATION RACK, 8 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, 1 Sided Bead Pellet Location, Low Position, 52MGO, NdFeB Magnet, 13mm Center Spacing, 10.9mm Hole Diameter, HDPE (High Density Polyethylene)
This item is not returnable. View return policy
Catalog No. NC2137756 Supplier V&P Scientific Inc Supplier No. VP 772F61
MAGNETIC BEAD SEPARATION RACK, 8 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, 1 Sided Bead Pellet Location, Low Position, 52MGO, NdFeB Magnet, 13mm Center Spacing, 10.9mm Hole Diameter, HDPE (High Density Polyethylene)