Learn More
V&P Scientific Inc MAGNETIC RACK, 8 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, Bottom Position
Catalog No. NC1460681
Encompass
MAGNETIC BEAD SEPARATION RACK, 8 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, Bottom Bead Pellet Location, Bottom Position, 52MGO, NdFeB Magnet, 13mm Center Spacing, 10.9mm Hole Diameter, HDPE (High Density Polyethylene)
Catalog No. NC1460681 Supplier V&P Scientific Inc Supplier No. VP 772F6
MAGNETIC BEAD SEPARATION RACK, 8 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, Bottom Bead Pellet Location, Bottom Position, 52MGO, NdFeB Magnet, 13mm Center Spacing, 10.9mm Hole Diameter, HDPE (High Density Polyethylene)