Learn More
Seracare Life Sciences Inc MAG BEADS ANTI-E.COLI O121 1ML
Catalog No. 50607728
Encompass_Preferred
Anti-E.coli O121 Magnetic Beads are super-paramagnetic polystyrene beads coated with KPL's Anti-E.coli O121 Antibody. They are intended for use in the isolation and separation of E.coli O121 from a variety of samples.
This item is not returnable. View return policy
Catalog No. 50-607-728 Supplier Seracare Life Sciences Inc Supplier No. 53500043
Anti-E.coli O121 Magnetic Beads are super-paramagnetic polystyrene beads coated with KPL's Anti-E.coli O121 Antibody. They are intended for use in the isolation and separation of E.coli O121 from a variety of samples.