Learn More
Seracare Life Sciences Inc MAG BEADS ANTI-E.COLI O157 1ML
Catalog No. 50607723
Encompass_Preferred
Anti-E.coli O157 Magnetic Beads are super-paramagnetic polystyrene beads coated with KPL's Anti-E.coli O157, Molecular Grade Antibody. They are intended for use in the isolation and separation of E.coli O157 from a variety of samples.
This item is not returnable. View return policy
Catalog No. 50-607-723 Supplier Seracare Life Sciences Inc Supplier No. 53500037
Anti-E.coli O157 Magnetic Beads are super-paramagnetic polystyrene beads coated with KPL's Anti-E.coli O157, Molecular Grade Antibody. They are intended for use in the isolation and separation of E.coli O157 from a variety of samples.