Learn More
Omega Biotek Inc Mag-Bind Blood & Tissue DNA HDQ 96 Kit, 4 x 96 preps, Magnetic Beads, Input Volume Up to 250 μL (Whole Blood), 50-200 μL Elution Volume, 8-384 samples throughput, No organic solvents
Catalog No. NC0555576
Encompass
The Mag-Bind Blood & Tissue DNA HDQ 96 Kit from OMEGA BIOTEK INC offers a rapid and efficient solution for isolating high-quality genomic DNA from various biological samples. This kit employs advanced magnetic bead technology to ensure excellent nucleic acid binding and purification, allowing researchers to achieve DNA that meets the demanding requirements of downstream applications. With its quick magnetic response and compatibility with numerous automation platforms, it is ideally suited for high-throughput laboratory environments. The purified DNA is of high molecular weight and free of any inhibitors, providing robust performance for applications such as NGS, qPCR, and PCR.
Catalog No. NC0555576 Supplier Omega Biotek Inc Supplier No. M639901
The Mag-Bind Blood & Tissue DNA HDQ 96 Kit from OMEGA BIOTEK INC offers a rapid and efficient solution for isolating high-quality genomic DNA from various biological samples. This kit employs advanced magnetic bead technology to ensure excellent nucleic acid binding and purification, allowing researchers to achieve DNA that meets the demanding requirements of downstream applications. With its quick magnetic response and compatibility with numerous automation platforms, it is ideally suited for high-throughput laboratory environments. The purified DNA is of high molecular weight and free of any inhibitors, providing robust performance for applications such as NGS, qPCR, and PCR.