Learn More
Omega Biotek Inc Mag-Bind Blood & Tissue DNA HDQ 96 Kit, 96-well plate, genomic DNA isolation, >60 kb DNA size, high-quality, inhibitor-free
Catalog No. NC2101454
Encompass
Mag-Bind Blood & Tissue DNA HDQ 96 Kit, 96-well plate, genomic DNA isolation, >60 kb DNA size, high-quality, inhibitor-free
This item is not returnable. View return policy
Catalog No. NC2101454 Supplier Omega Biotek Inc Supplier No. M639900
Mag-Bind Blood & Tissue DNA HDQ 96 Kit, 96-well plate, genomic DNA isolation, >60 kb DNA size, high-quality, inhibitor-free