Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omega Biotek Inc Mag-Bind SeqDTR, Magnetic Bead-based purification system, 500 mL, Manual or automated, 96-well, 384-well, Over 800 bps

Catalog No. NC2253341
Encompass

The Omega Bio-tek Mag-Bind SeqDTR is a magnetic bead-based purification system designed for the efficient and reliable removal of unincorporated dye terminators from sequencing reactions. This product employs proprietary chemistry and utilizes the unique properties of paramagnetic beads, enhancing sample purity and quality in sequencing applications. It is suitable for both manual and automated workflows, allowing users to achieve optimal performance with ease.

  • Application: Removal of unincorporated dye terminators from sequencing reactions
  • Technology: Magnetic bead based
  • Kit volume: 500 mL
  • Automation compatibility: Manual or automated
  • Average read lengths: Over 800 bps (Min Phred 20)
  • Reduce reagent usage: 1/16 or 1/32 compared to sephadex cleanups
  • Contaminants removed: Nucleotides, primers, salts, dye terminators
  • Storage conditions: 2-8 °C
  • Processing time: Less than 25 minutes for 4 plates

This item is not returnable. View return policy
Catalog No. NC2253341 Supplier Omega Biotek Inc Supplier No. M130050
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Omega Bio-tek Mag-Bind SeqDTR is a magnetic bead-based purification system designed for the efficient and reliable removal of unincorporated dye terminators from sequencing reactions. This product employs proprietary chemistry and utilizes the unique properties of paramagnetic beads, enhancing sample purity and quality in sequencing applications. It is suitable for both manual and automated workflows, allowing users to achieve optimal performance with ease.

  • Application: Removal of unincorporated dye terminators from sequencing reactions
  • Technology: Magnetic bead based
  • Kit volume: 500 mL
  • Automation compatibility: Manual or automated
  • Average read lengths: Over 800 bps (Min Phred 20)
  • Reduce reagent usage: 1/16 or 1/32 compared to sephadex cleanups
  • Contaminants removed: Nucleotides, primers, salts, dye terminators
  • Storage conditions: 2-8 °C
  • Processing time: Less than 25 minutes for 4 plates

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.