Learn More
Omega Biotek Inc Mag-Bind TotalPure NGS, cleanup and size selection, 50 mL volume, >100 bp binding size, DNA/RNA compatible, typical recovery rate 80%, double/single-sided size selection
Catalog No. NC1545573
Encompass
Mag-Bind TotalPure NGS, cleanup and size selection, 50 mL volume, >100 bp binding size, DNA/RNA compatible, typical recovery rate 80%, double/single-sided size selection
Catalog No. NC1545573 Supplier Omega Biotek Inc Supplier No. M137801
Mag-Bind TotalPure NGS, cleanup and size selection, 50 mL volume, >100 bp binding size, DNA/RNA compatible, typical recovery rate 80%, double/single-sided size selection