Learn More
Omega Biotek Inc Mag-Bind Total RNA 96 Kit | 4 x 96 preps
Catalog No. NC1715458
Encompass
The Mag-Bind Total RNA 96 Kit provides a novel technology for total RNA isolation. This kit allows the rapid and reliable isolation of high-quality total cellular RNA and viral RNA from a wide variety of cells and tissues. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which improves the removal of contaminants and increases nucleic acid purity. Mag-Bind Total RNA 96 Kit procedure can be fully automated with most robotic workstations.
Catalog No. NC1715458 Supplier Omega Biotek Inc Supplier No. M673100
The Mag-Bind Total RNA 96 Kit provides a novel technology for total RNA isolation. This kit allows the rapid and reliable isolation of high-quality total cellular RNA and viral RNA from a wide variety of cells and tissues. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which improves the removal of contaminants and increases nucleic acid purity. Mag-Bind Total RNA 96 Kit procedure can be fully automated with most robotic workstations.