Learn More
Omega Biotek Inc NGS Workflow PCR Clean Up Mag Bind Total Pure NGS
Catalog No. NC1990887
Encompass
The Mag-Bind TotalPure NGS kit efficiently purifies PCR products for next-generation sequencing, removing impurities with high recovery rates. It includes all necessary components for processing 5-100μL PCR volumes and is compatible with major sequencing platforms.
Catalog No. NC1990887 Supplier Omega Biotek Inc Supplier No. M137800
The Mag-Bind TotalPure NGS kit efficiently purifies PCR products for next-generation sequencing, removing impurities with high recovery rates. It includes all necessary components for processing 5-100μL PCR volumes and is compatible with major sequencing platforms.