Learn More
Omega Biotek Inc Ultra-Pure Plasmid DNA 96 Kit, 4 x 96 preps
Catalog No. NC1143575
Encompass
Ultra-Pure Plasmid DNA 96 Kit combines the power of Mag-Bind technology with the innovative ETR technology (Endo-toxin removal) to deliver high-quality endotoxin-free plasmid DNA in high throughput format.
Catalog No. NC1143575 Supplier Omega Biotek Inc Supplier No. M125801
Ultra-Pure Plasmid DNA 96 Kit combines the power of Mag-Bind technology with the innovative ETR technology (Endo-toxin removal) to deliver high-quality endotoxin-free plasmid DNA in high throughput format.