Learn More
ACROBiosystems ACROBiosystems Human MAG / Siglec-4a Protein, Fc Tag
Supplier:  ACROBiosystems MAGH52541MG
Encompass_Preferred
Recombinant Protein;1MG;Human MAG, Fc Tag (MAG-H5254) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Pro 516 (Accession # P20916-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.1 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6663