Learn More
Aat Bioquest Mag-Fluo-4 AM (10x50 ug)
SDP
Catalog No. 502249827
Encompass_Preferred
The cell-permeant Mag-Fluo-4 AM is an analog of Fluo-4 AM with a Kd for Mg ion of 4.7 mM and a Kd for Ca ion of 22 microM making it useful as an intracellular Mg ion indicator as well as a low-affinity Ca ion indicator. This catalog is for 10x50 ug.
This item is not returnable. View return policy
Catalog No. 50-224-9827 Supplier Aat Bioquest Supplier No. 20401
Add to Cart
Edge
Add to Cart
The cell-permeant Mag-Fluo-4 AM is an analog of Fluo-4 AM with a Kd for Mg ion of 4.7 mM and a Kd for Ca ion of 22 microM making it useful as an intracellular Mg ion indicator as well as a low-affinity Ca ion indicator. This catalog is for 10x50 ug.