Learn More
Aat Bioquest Mag-Fura-2 AM Cell-permeant (10x50 ug)
SDP
Catalog No. 502249825
Encompass_Preferred
Mag-Fura-2 AM is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This catalog is for 10x50 ug.
This item is not returnable. View return policy
Catalog No. 50-224-9825 Supplier Aat Bioquest Supplier No. 20383
Mag-Fura-2 AM is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This catalog is for 10x50 ug.