Learn More
Biotium Furaptra, AM Ester, 20x50ug
Catalog No. 501964697
Encompass_Preferred
Furaptra, AM ester is a membrane-permeant form of Furaptra (Mag-Fura-2). The indicator is loaded into cells via simple incubation, where it is cleaved by cytoplasmic esterases to release Mag-Fura-2. Properties: Light yellow solid soluble in DMSO; Store at -20°C and protect from light; C30H30N2O19; MW: 723
Catalog No. 50-196-4697 Supplier Biotium Supplier No. 50039
Furaptra, AM ester is a membrane-permeant form of Furaptra (Mag-Fura-2). The indicator is loaded into cells via simple incubation, where it is cleaved by cytoplasmic esterases to release Mag-Fura-2. Properties: Light yellow solid soluble in DMSO; Store at -20°C and protect from light; C30H30N2O19; MW: 723