Learn More
Aat Bioquest Mag-Fura-2 tetrapotassium salt Cell impermeant (1 mg)
SDP
Catalog No. 502249824
Encompass_Preferred
Mag-Fura-2 tetrapotassium salt is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9824 Supplier Aat Bioquest Supplier No. 20380
Mag-Fura-2 tetrapotassium salt is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This catalog is for 1 mg.