Learn More
ACROBiosystems ACROBiosystems Human MAG / Siglec-4a Protein, His Tag
Catalog No. 501956659
Encompass_Preferred
Recombinant Protein;1MG;Human MAG, His Tag (MAG-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Pro 516 (Accession # P20916-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6659 Supplier ACROBiosystems Supplier No. MAGH52H81MG
Recombinant Protein;1MG;Human MAG, His Tag (MAG-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Pro 516 (Accession # P20916-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.