Learn More
Ibidi USA Magnetic Lid for µ-Dish Microscopy Rack
SDP
Catalog No. 50305730
Encompass
Lid fixed with magnetic positioning for additional stabilization. Compatible with the µ-Dish Microscopy Rack. Includes 1 lid for µ-Dish 35mm, High. aluminum, black, Ø 51 x 10.95 mm
This item is not returnable. View return policy
Catalog No. 50-305-730 Supplier Ibidi USA Supplier No. 80037
Lid fixed with magnetic positioning for additional stabilization. Compatible with the µ-Dish Microscopy Rack. Includes 1 lid for µ-Dish 35mm, High. aluminum, black, Ø 51 x 10.95 mm