Learn More
MSC Mag-Mate Ceramic Magnetic Grate Separator, 10 Inch Square, 5 Tubes
Catalog No. 5000531036
Encompass
This ceramic magnetic grate separator is designed for effective and economical ferrous tramp metal separation. It is ideal for use in various applications including bins, chutes, drawers, hoppers, and circular enclosures, providing an economical solution for capturing and separating large pieces of unwanted steel from products.
  • Constructed with 1 inch diameter magnetic tubes placed on 2 inch centers.
  • Features heavy-duty 1/8 inch thick by 2 inch wide stainless steel framing.
  • Includes 2GA thick by 2 inch wide stainless steel support stringers.
  • Custom sizes and designs available.
  • Sanitary or food-grade construction options.
  • Product diverter options.
  • Custom tube spacing options.
  • Coatings available for abrasive products.
Catalog No. 50-005-31036 Supplier MSC Supplier No. 64830466
This ceramic magnetic grate separator is designed for effective and economical ferrous tramp metal separation. It is ideal for use in various applications including bins, chutes, drawers, hoppers, and circular enclosures, providing an economical solution for capturing and separating large pieces of unwanted steel from products.
  • Constructed with 1 inch diameter magnetic tubes placed on 2 inch centers.
  • Features heavy-duty 1/8 inch thick by 2 inch wide stainless steel framing.
  • Includes 2GA thick by 2 inch wide stainless steel support stringers.
  • Custom sizes and designs available.
  • Sanitary or food-grade construction options.
  • Product diverter options.
  • Custom tube spacing options.
  • Coatings available for abrasive products.