Learn More
Grainger MAG-MATE DISC MAGNET
Catalog No. NC2237024
Encompass_Preferred
MAG-MATE Disc Magnet: Grade 35 Neodymium, Nickel Plating, 44 lb Max. Pull, 1/2 in Overall Lg
This item is not returnable. View return policy
Catalog No. NC2237024 Supplier Grainger Supplier No. 31LU59
MAG-MATE Disc Magnet: Grade 35 Neodymium, Nickel Plating, 44 lb Max. Pull, 1/2 in Overall Lg