Learn More
MSC Mag-Mate Magnetic Grate Rod Separator, 1 in Diameter, 4 in Length, Ceramic Magnet
Catalog No. 5000535994
Encompass
This magnetic grate rod separator is designed to efficiently remove ferrous contaminants from product flow. Featuring a durable stainless steel rod and ceramic magnet, it provides reliable magnetic separation in various industrial applications. The 1 in diameter and 4 in length make it suitable for compact spaces, while the tapped end plugs allow for easy installation.
  • Removes ferrous contaminants from product flow.
  • Durable stainless steel rod.
  • Ceramic magnet for effective separation.
  • Tapped end plug mounting holes (1/4-20) for easy bolt-in installation.
  • Rolled ends with stainless steel plugs.
  • Available in various standard and custom lengths.
Catalog No. 50-005-35994 Supplier MSC Supplier No. 64830268
This magnetic grate rod separator is designed to efficiently remove ferrous contaminants from product flow. Featuring a durable stainless steel rod and ceramic magnet, it provides reliable magnetic separation in various industrial applications. The 1 in diameter and 4 in length make it suitable for compact spaces, while the tapped end plugs allow for easy installation.
  • Removes ferrous contaminants from product flow.
  • Durable stainless steel rod.
  • Ceramic magnet for effective separation.
  • Tapped end plug mounting holes (1/4-20) for easy bolt-in installation.
  • Rolled ends with stainless steel plugs.
  • Available in various standard and custom lengths.